Precision balance, ME4002/M
Lieferant: Mettler - Toledo
30046451
METB30046451EA 2855 EUR
METB30046451
Precision balance, ME4002/M
Waagen
State-of-the art weighing technology, solid construction and high quality materials ensure the accurate results.
  • Hygenic design
  • Easy, efficient and functional
  • Backlit LCD
  • Maximum capacity: 4,2 kg
The easy-to-use interface enables direct access to applications and also the unit is coupled with easy cleaning to ensures the fast and efficient action. The large numbers on the brilliant backlit display are easy to read in all working environments. With the built-in function, the date and time can be added to the results in accordance with ISO/GLP documentation requirements.