OptoCap for optical DO sensor
Lieferant: Mettler - Toledo
30018857
METN30018857EA 655 EUR
METN30018857
OptoCap for optical DO sensor
Elektroden Sauerstoff-Elektroden
The oxygen-sensitive OptoCap for the InPro 6860i sensor requires only a few seconds to replace.
  • PTFE layer avoids cell accumulation
  • Electropolished surface
  • EPDM O-ring
  • Autoclavable
Durable materials, robust design and easy replacement. Exchange of the OptoCap is recommended after a maximum 360 days. Exchange ensures high accuracy and performance over the sensor's lifetime.
Standards: Certified to FDA/USP class VI, 3.1, N5/Ra 16.