Titrators, Karl Fisher, EasyPlus™
Lieferant: Mettler - Toledo
30060041 30060045
METN30060041EA 3645 EUR
METN30060041 METT30060045
Titrators, Karl Fisher, EasyPlus™
Titratoren
This versatile titrator is suitable for a range of determinations making it ideal for simple titrations of most sample types. The simple 'App' style user interface reduces training time while increasing work speed and simplicity. iTitrate Intelligence’s guided set-up assists in method programming to ensure samples are tested consistently, regardless of operator, and with minimal effort. Ideal for KF titrations of a wide range of samples from any industry.
  • Very small footprint
  • LongClick™ function starts routine analysis from the home screen
  • iTitrate Intelligence enables very simpe method programming with EasySetup guidance; titrator automatically calculates results using predefined formulas
  • Injection port for syringes and pipetted liquid samples
  • Handy weighing/carrying spoon for easy and spill-free solid sample introduction
Easy pH: Ideal for automated acid/base titrations including the determination of acidity in juices, milk and wine plus p- and m-values in water and much more.
Easy Cl: Ideal for chloride determination in food and other products, plus silver content determination by precipitation.
Easy Pro: Combining the three main types of titrations: acid/base, precipitation and redox, this instrument determines the most common parameters in food, beverage and chemical products.
Easy KFV: Ideal for routine volumetric Karl Fischer titration from 100 ppm up to 100% in food, chemical products and other sample types.