Cup lid, DP
Lieferant: Mettler - Toledo
51143031
METN51143031EA 16 EUR
METN51143031
Cup lid, DP
Deckel Lids for Laboratory Vessels/Containers
Safe and clean operation.
Closed cup lid with vent hole prevents spillage(contamination) in the event of expanding sample.