Karl Fisher drying oven, DO308
Lieferant: Mettler - Toledo
51371200
METN51371200EA 0 EUR
METN51371200
Karl Fisher drying oven, DO308
Öfen Vakuumtrockenschränke
The DO308 drying oven is the ideal accessory for the METTLER TOLEDO volumetric or coulometric Karl Fischer titrators. It allows easy and accurate water determination of solid or viscous samples.
  • Easy handling for left and right handed operators
  • Sample exchange and sample transport into the heated zone of the oven is provided for by the simple transport system with a magnetic guide rod attached to the glass sample boat
  • Equilibrium conditions are quickly reached: Cold and the hot zone can be purged separately with dry carrier gas using a stopcock
  • Ambient moisture introduced during sample exchange or addition via the sleeve opening is quickly carried away; therefore, a stable drift is reached within a short period of time
  • There is no need to remove the transfer tube from the titration cell after switching off the oven; just open the stopcock to avoid contamination of the oven due to underpressure
  • Different temperature ranges can be pre-programmed in one cycle
  • Automatic shut down after the analysis is completed
  • Ready for use: All necessary parts including the drying unit with gas flow measurement are part of the standard delivery
The DO308 drying oven is ideal for accurate water determination of solid or viscous samples like:
- Substances that release water only at elevated temperatures such as plastics in powder or granular form
- Substances that cause side reactions with Karl Fischer reagent such as oxidizing inorganic salts
- Substances that cause problems in the titration vessel due to their consistency such as fibrous or pasty substances
- Substances that are poorly soluble or insoluble such as wool, dough, tar or coal
Bestellinformation: If purging with dry air is desired, an optional air pump is available.
Lieferumfang: Supplied with: Gas drying unit including flow meter, Glass sample boat (2x), Sample boat guide rod, Set of 50 aluminium inserts for the sample boat, Tube Set, Stop cock and Transfer tube (insulated).