pH Sensor, InPro3250i SG/325
Lieferant: Mettler - Toledo
52005375
METN52005375EA 879 EUR
METN52005375
pH Sensor, InPro3250i SG/325
Elektroden pH-Elektroden
The InPro 3250i SG/325 is a digital, pre-pressurized, low maintenance ISM pH and ORP sensor combined with a temperature sensor for in-line measurement in demanding applications. The 325 mm version is optimal for medium to long installation lengths.
  • High performance, high accuracy pH and ORP
  • Highly accurate in extreme conditions and easy validation
  • Low maintenance and easy-to-use
  • Sterilizable and autoclavable
  • Reliable, accurate and robust
  • Advanced sensor diagnostics with ISM
This sensor can measure both pH and ORP (Redox), and has a built in digital temperature sensor. It is pressure resistant up to 4 barg (58 psig), has a watertight connector and can withstand repeated sterilization and autoclave cycles.
The InPro 3250i SG/325 provides stable and precise readings under extreme process conditions from pH 0 to 14 with its high-alkali pH glass membrane.
Standards: FM: IS Cl. I, II, III, Div 1, GR ABCDEFG/T6 EHEDG; ATEX/IECEx: Ex ia IIC T6/T5/T4/T3 Ga/Gb PED.