pH/mV/°C meters, handheld, Calimatic® 1120/1140
Lieferant: Mettler - Toledo
52120657
METN52120657EA 0 EUR
METN52120657
pH/mV/°C meters, handheld, Calimatic® 1120/1140
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
The innovative design of these pH meters allow optimum use of the meters in a whole variety of applications. Suitable for laboratory and industrial-scale applications, e.g. cheese-making, slaughter-houses (support for HACCP processes), breweries, spot-check measurements, as well as for all kinds of field use.
  • Well-arranged chemically resistant foil keypad makes the instrument easy to operate and highly user-friendly
  • Easy-to-read LC display
  • Ergonomic design
  • Calimatic system automatically carries out all necessary unit functions for calibration of the sensor, including single-point or dual-point calibration as selected
  • Sensoface feature provides information on the condition of the sensor by displaying appropriate "smilies"
The explosion-proof version 1120-X has been specially designed for use in potentially explosive areas in the chemical industry. EEX ia IIC T6 and FM Class I Div I.
Lieferumfang: Supplied with carrying strap, batteries, instruction manual and short instructions.