Shelving systems, stem castor cart with solid shelves, Super Erecta®
Lieferant: Metro International
F556EG
METOF556EGEA 1790 EUR
METOF556EG
Shelving systems, stem castor cart with solid shelves, Super Erecta®
Möbel Regale
The system consists of four solid shelves, posts, plastic split sleeves, donut bumpers and 127 mm poly castors (2 swivel; 2 brake). The raised 'ship's edge' around perimeter helps to contain spillage.
  • Chrome-plated posts with four galvanised flat shelves