Super Erecta® 4-Shelf Industrial Wire Shelving Starter Unit
Lieferant: Metro International
SE-246074K4-S4
METOSE-246074K4-S4EA 0 EUR
METOSE-246074K4-S4
Super Erecta® 4-Shelf Industrial Wire Shelving Starter Unit
Möbel Regale
Super Erecta® Metroseal gray shelving is protected by a layer of rust-resistant epoxy laid over a protective zinc substrate. These corrosion-resistant shelving units are NSF-approved for use in wet environments.
Metroseal Gray shelving combines aesthetics and function into one, cross-industry solution. This 4-tier shelf is a clean, safe storage solution designed for use in the foodservice, healthcare, and laboratory industries. Each unit is easy to assemble with minimal tools and can be adjusted by 1" increments on Metro SiteSelect posts.
Each Metroseal unit contains Microban antimicrobial protection to inhibit the growth of mold, mildew, fungi, and bacteria. On an unprotected surface, stain and odor-causing bacteria can double in number every 20 minutes. Microban protection works continuously to keep products cleaner for longer. Each 4-tier shelving kit comes complete with 4 shelves with split sleeves and 4 posts with leveling bolts.