Titrators, coulometric, 756/831 KF
Lieferant: METROHM
2.831.0010
METR2.831.0010EA 13590 EUR
METR2.831.0010
Titrators, coulometric, 756/831 KF
Titratoren
The 756 and 831 coulometers are ideal for water content determination down to trace levels with guaranteed precision and accuracy.
  • Backlit graphical display, multi-language dialog, easy to use
  • Screen provides a clear presentation of the parameters and graphically shows the course of the KF determination (live curve, μg water versus time)
  • Two RS-232 interfaces allow communication with a balance, external printer and/or computer
  • Standard and Expert mode: In the standard mode only those functions are accessible that are needed in routine work
  • Integrated method memory has space for about 100 methods
  • Can be operated together with a KF oven
Recommended measuring range: 10 µg - 200 mg (absolute water)
Bestellinformation: Supplied with generator electrode and comprehensive accessories.