Magnetic Stirrer with Stand
Lieferant: METROHM
28010040
METR28010040EA 0 EUR
METR28010040
Magnetic Stirrer with Stand
Rührer Magnetrührer
Magnetic stirrer including base plate, support rod and electrode holder for supplementing the Titrino plus, Dosimat plus, titrandos, sample processors, 805 Dosimat and 780/781 pH meters as well as the 856 and 867 measuring modules.
With permanently attached cable for MSB (Metrohm Serial Bus).