Septum caps
Lieferant: METROHM
6.1448.057
METR6.1448.057EA 691 EUR
METR6.1448.057
Septum caps
Verschlüsse Chromatographieampullen-Verschlüsse
Aluminium caps with silicone septum suitable for 6 ml sample vials.