RS-232 Cable
Lieferant: METROHM
6.2125.110
METR6.2125.110EA 119 EUR
METR6.2125.110
RS-232 Cable
Kabel
Connecting RS-232 interface to PC. Used with pH meters, titrinos, KF coulometers, VA trace analyser.