pH buffer solutions, NIST/DIN
Lieferant: Mettler - Toledo
51350052
51350054
51350058
51350056
30111136
30111137
30111138
30111139
METT51350052EA
28.5
EUR
METT51350052
METT51350054
METT51350058
METT51350056
METT30111136
METT30111137
METT30111138
METT30111139
pH buffer solutions, NIST/DIN
Puffer
pH-Puffer
The NIST/DIN pH buffers are specified to three decimal places, e.g. 9.180, offering the highest possible pH precision.
A detailed test certificate is available for every bottle and guarantees the tested values and traceability.
Standards: NIST traceable.