pH buffer solutions, NIST/DIN
Lieferant: Mettler - Toledo
51350052 51350054 51350058 51350056 30111136 30111137 30111138 30111139
METT51350052EA 28.5 EUR
METT51350052 METT51350054 METT51350058 METT51350056 METT30111136 METT30111137 METT30111138 METT30111139
pH buffer solutions, NIST/DIN
Puffer pH-Puffer
The NIST/DIN pH buffers are specified to three decimal places, e.g. 9.180, offering the highest possible pH precision.
A detailed test certificate is available for every bottle and guarantees the tested values and traceability.
Standards: NIST traceable.