Reference electrode filling solutions
Lieferant: Mettler - Toledo
51343185
51343182
51343184
51343180
51350084
51350076
51350074
51350072
51350082
662-2886EA
12
EUR
662-2886
METT51343182
METT51343184
METT51343180
METT51350084
METT51350076
METT51350074
METT51350072
METT51350082
Reference electrode filling solutions
Elektrolyte
Elektrodenfülllösungen
FRISCOLYT-B® (84,4% glycerol, 11% KCl and 4,6% H₂O) is for low temperature samples.