pH buffer solutions, NIST/DIN
Lieferant: Mettler - Toledo
The NIST/DIN pH buffers are specified to three decimal places, e.g. 9.180, offering the highest possible pH precision.
A detailed test certificate is available for every bottle and guarantees the tested values and traceability.
Standards: NIST traceable.