pH combination electrode, open junction, InLab®
Lieferant: Mettler - Toledo
52000379
METT52000379EA 335 EUR
METT52000379
pH combination electrode, open junction, InLab®
Elektroden pH-Elektroden
Ideal for a wide range of applications including emulsions, suspensions, effluent water, and solutions containing proteins and sulfides. Electrode meets IP 67 specifications, allowing the sensor to be immersed in water to a depth of 1 m for one hour without any water incursion.
  • With integrated temperature sensor, fixed cable, and BNC/Cinch connector
  • Solid polymeric electrolyte requires no junction at all
  • High-grade plastic (PEEK™) shaft offers high level of chemical resistance and robustness
  • ARGENTHAL™ reference system improves service life and measurement accuracy at differing sample temperatures
Lieferumfang: Supplied with quality certificate.