Polypropylene and PVC Pressure Regulators
Lieferant: Avantor Fluid Handling
01350-82
MFLX01350-82EA 894 EUR
MFLX01350-82
Polypropylene and PVC Pressure Regulators
Ventile Überdruckventile
Easily control your outlet pressure.
  • Choose between PVC or polypropylene (PP) construction with Viton® diaphragm
Valves feature a diaphragm made of Viton®. Use to control your outlet pressure between 5 and 50 psig; up to 150 psig inlet. Ideal for your liquid applications.