Hedland Spring-Loaded In-Line Flowmeters
Lieferant: Avantor Fluid Handling
03231-23 03231-27 03231-29 03231-01 03231-03 03231-33
MFLX03231-23EA 200 EUR
MFLX03231-23 MFLX03231-27 MFLX03231-29 MFLX03231-01 MFLX03231-03 MFLX03231-33
Hedland Spring-Loaded In-Line Flowmeters
Durchflussmesser
Provide accurate readings while mounted in any position.
  • Rugged design resists shock and vibration
  • Control pumps or valves with an optional limit switch
These economical flowmeters are built with a spring-retained piston for direct, accurate readings regardless of the position you mount them. Use them to monitor hydraulic systems or chemical processes; the meters also work well to check pump and control valve performance.
Note: Polysulfone is not suited for direct contact with sunlight.
Considering limit switches? Use to provide an output signal at a setpoint anywhere between 0 to 100% of full-scale. Use to control small pumps, valves, or alarms or to interface with a PLC.