Bürkle Thread Adapters for Hand-Operated Mini Solvent Pump
Lieferant: Avantor Fluid Handling
06040-02
MFLX06040-02EA 59.3 EUR
MFLX06040-02
Bürkle Thread Adapters for Hand-Operated Mini Solvent Pump
Pumpen Fass/Transferpumpen
Suitable for use with Hand-Operated Mini Solvent Pumps.
  • Order optional adapters to fit a wide range of containers
  • Avaiable in different material and thread configurations
  • Withstand temperatures up to 104 °F (40 °C)
The 06043-95 hand pump features a GL45 connecting thread made of PTFE for connection to a wide range of containers. Optional S40, GL38 and GL32 adapters in PTFE or ETFE are available separately to easily adapt this hand operating pump to other bottles, canisters, or tanks.