Bürkle Hand-Operated Pump Dispensers for Commercial Storage Drums
Lieferant: Avantor Fluid Handling
06040-45 06040-46 06040-53
MFLX06040-45EA 110 EUR
MFLX06040-45 MFLX06040-46 MFLX06040-53
Bürkle Hand-Operated Pump Dispensers for Commercial Storage Drums
Pumpen Fass/Transferpumpen
Dispense liquids safely and quickly from most storage containers.
  • Telescopic immersion tube adapts itself to a variety of containers
  • Different materials and styles suit a wide range of applications
These hand-operated pumps feature a universal soft PVC bung consisting of a conical center and two conical sleeves allowing for an airtight seal between the pump and most liquid storage container openings. The telescopic immersion tube adapts itself to the container height on insertion, ensuring good removal of residual liquid. Various tube diameters are available to meet your individual liquid transfer needs. Choose from polypropylene (PP), PVDF, or 304 stainless steel (SS) wetted materials and dispensing by spout or hose.