Hand-Operated Pump Dispensers for Barrels and Canisters
Lieferant: Avantor Fluid Handling
06043-93
MFLX06043-93EA 855 EUR
MFLX06043-93
Hand-Operated Pump Dispensers for Barrels and Canisters
Pumpen Fass/Transferpumpen
Ideal for contamination-free removal of organic solvents.
  • Designed for use with 15.8 gal. (60 L) containers
  • Sturdy stainless steel and PTFE construction ensures long life
  • Suitable for containers with R 2” thread or pull-out spout
The hand-operated solvent pumps, with a capacity of 10 L per minute, safely dispense organic solvents and flavoring agents. Two ball valves prevent dangerous vapors from escaping after dispensing by providing a gas-tight, completely sealable pump. Model 06043-91 is ideal for stainless steel barrels with a R 2” thread, while model 06043-93 is suitable for tin-foil canisters with a pull-out spout. Order optional antistatic grounding set for pumping flammable liquids.
Lieferumfang: Includes thread adapter for 06043-93.