Conical Universal Barbed Straight Connector, PVDF
Lieferant: Avantor Fluid Handling
06044-07
MFLX06044-07EA 50.8 EUR
MFLX06044-07
Conical Universal Barbed Straight Connector, PVDF
Fittings und Verbinder
Safe and flexible connection for your most demanding applications.
  • Withstands temperatures up to 320 °F (160 °C)
  • Highly resistant to chemicals and UV
  • Autoclavable
The conical universal barbed straight connector is ideal for extending, connecting, branching, joining, and repairing most plastic tubings. The polyvinylidene fluoride (PVDF) connector are designed for quick and easy assembly without any tools.