CPC® Quick-Disconnect Fittings, Hose Barb Bodies
Lieferant: Avantor Fluid Handling
06360-09 06360-10 06360-12 06360-15 06360-19 06360-20 06360-22 06360-25 06360-55 06360-57 06361-03 06361-04 06361-05 06364-09 06364-10 06364-15 06364-19 06364-55 06364-60 06361-04BLK 06361-03BLK 06360-10BLK 06360-12BLK 06360-15BLK 06360-55BLK 06364-09BLK 06364-10BLK 06364-55BLK 06360-09BLK 06361-05BLK 06364-15BLK 06364-60BLK 06360-57BLK 06364-19BLK 06360-25BLK 06360-19BLK 06360-20BLK 06360-22BLK
MFLX06360-60EA 26.2 EUR
MFLX06360-60 MFLX06360-65 MFLX06360-67 MFLX06360-70 MFLX06361-11 MFLX06361-12 MFLX06361-46 MFLX06361-48 MFLX06361-51 MFLX06361-53 MFLX06364-20 MFLX06364-25 MFLX06364-65 MFLX06364-70 MFLX06360-09 MFLX06360-10 MFLX06360-12 MFLX06360-15 MFLX06360-19 MFLX06360-20 MFLX06360-22 MFLX06360-25 MFLX06360-55 MFLX06360-57 MFLX06361-03 MFLX06361-04 MFLX06361-05 MFLX06364-09 MFLX06364-10 MFLX06364-15 MFLX06364-19 MFLX06364-55 MFLX06364-60 MFLX06361-04BLK MFLX06361-03BLK MFLX06360-10BLK MFLX06360-12BLK MFLX06360-15BLK MFLX06360-55BLK MFLX06364-09BLK MFLX06364-10BLK MFLX06364-55BLK MFLX06360-09BLK MFLX06361-05BLK MFLX06364-15BLK MFLX06364-60BLK MFLX06360-57BLK MFLX06364-19BLK MFLX06360-25BLK MFLX06360-19BLK MFLX06360-20BLK MFLX06360-22BLK MFLX06361-46BLK MFLX06361-51BLK MFLX06361-12BLK MFLX06361-53BLK MFLX06360-60BLK MFLX06361-11BLK MFLX06364-25BLK MFLX06364-65BLK MFLX06364-70BLK MFLX06361-48BLK MFLX06360-65BLK MFLX06360-67BLK MFLX06360-70BLK MFLX06364-20BLK MFLX06361-52BLK
CPC® Quick-Disconnect Fittings, Hose Barb Bodies
Fittings und Verbinder
No more struggling - just press thumb latch.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).