CPC® Quick-Disconnect Fittings, Hose Barb Elbow Inserts
Lieferant: Avantor Fluid Handling
06362-01 06362-01BLK 31307-67BLK 31307-68BLK
MFLX06361-27EA 9.4 EUR
MFLX06361-27 MFLX06361-28 MFLX06361-39 MFLX06361-41 MFLX06361-63 MFLX06361-75 MFLX06361-76 MFLX06361-87 MFLX06361-98 MFLX06361-99 MFLX06362-00 MFLX06362-01 MFLX06362-01BLK MFLX06361-27BLK MFLX06361-41BLK MFLX06361-75BLK MFLX06361-76BLK MFLX06361-99BLK MFLX06361-28BLK MFLX06361-39BLK MFLX06361-87BLK MFLX06361-63BLK MFLX06361-98BLK MFLX06362-00BLK MFLX06361-64BLK MFLX31307-67BLK MFLX31307-68BLK
CPC® Quick-Disconnect Fittings, Hose Barb Elbow Inserts
Fittings und Verbinder
No more struggling - just press thumb latch.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).