Parker Hannifin Compression Fitting Inserts for Soft Tubing
Lieferant: Avantor Fluid Handling
06391-30
MFLX06391-30EA 9.1 EUR
MFLX06391-30
Parker Hannifin Compression Fitting Inserts for Soft Tubing
Fittings und Verbinder
Allows compression fittings to be used with soft tubing.
  • Secure your flexible, soft tubing
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Inserts fit in the end of the soft tubing to provide the rigidity required for adequate compression and a proper seal.