Multi-Channel Tubing Racks, Non-Adhesive
Lieferant: Avantor Fluid Handling
MFLX06432-03EA 25.5 EUR
MFLX06432-03 MFLX06432-04 MFLX06432-05 MFLX06432-08
Multi-Channel Tubing Racks, Non-Adhesive
Klemmen Schlauchklemmen und Sanitärklemmen
An easy solution to secure and route multiple tubing lines.
  • Keep your tubing lines organised and untangled
These tube racks offer a simple and economical method for organising tubing lines. The ³/₁₆" and ¹/₄" sizes can be easily separated down to single holders by hand for increased flexibility and custom tubing arrangements. The ³/₁₆" and ¹/₄" sizes are available with adhesive fastener, and all sizes are available with through holes for secure mounting (mounting screws are not included). Standard racks are black polyethylene; ³/₁₆" and ¹/₄" sizes areMore Product Information