Bev-A-Line® IV Transfer Pump Tubing
Lieferant: Avantor Fluid Handling
06490-12 06490-15 06490-17 06490-19 06490-20 06490-40
MFLX06490-12EA 44.5 EUR
MFLX06490-12 MFLX06490-15 MFLX06490-17 MFLX06490-19 MFLX06490-20 MFLX06490-40
Bev-A-Line® IV Transfer Pump Tubing
Schläuche Transferschlauch
Nontoxic and nonhemolytic.
  • Ideal for general-purpose, single-use, and extended fluid path applications
Applications: Safe for food, beverage, drug, lab, and general use—yet much more affordable than PTFE tubing. Safe for most alcohols; avoid use with strong solvents.
Characteristics: Polyethylene liner with ethyl vinyl acetate shell. No plasticizers. Nontoxic and nonhemolytic. Heat-bondable. Translucent.
Certification: Resin is FDA-compliant (liner: 21 CFR 177.1520; shell 21 CFR 177.1350). Meets USP Class VI, USDA, and National Formulary requirements.
More Product Information