Union Fitting, Male to Female Threaded Adapter, Elbow
Lieferant: Avantor Fluid Handling
06546-13 06546-14 06546-16
MFLX06546-13EA 78 EUR
MFLX06546-13 MFLX06546-14 MFLX06546-16
Union Fitting, Male to Female Threaded Adapter, Elbow
Fittings und Verbinder
Maintain a permanent connection.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Sturdy threaded adapters are designed for permanent installation. Choose the size and material that fits your application needs.