FMI Single-Head, Fixed-Speed AC Drives
Lieferant: Avantor Fluid Handling
07104-40 07104-10
MFLX07104-40EA 442 EUR
MFLX07104-40 MFLX07104-10
FMI Single-Head, Fixed-Speed AC Drives
Pumpen Pumpenantriebe Antrieb für Kolbenpumpen
No valves to clog or wear out.
  • Build your own pump systems using interchangeable heads, drives, and accessories
Series Q drives are interchangeable with Series Q pump heads. These fixed-speed AC drives feature a TEFC motor. Pedestal drive model 07104-40 is for chain-, belt-, or shaft-coupling to rotating machine elements or to your motor. Build your own pump systems using interchangeable heads, drives, and accessories. Pump heads sold separately.