Noshok® Metal Mini Needle Valves
Lieferant: Avantor Fluid Handling
08611-00 08611-02 08611-20 08611-26 08612-04
MFLX08611-00EA 54.8 EUR
MFLX08611-00 MFLX08611-02 MFLX08611-20 MFLX08611-26 MFLX08612-04
Noshok® Metal Mini Needle Valves
Ventile Nadelventile
Designed for industrial machinery, hydraulic, and pneumatic systems.
  • Viton® O-ring seal and PTFE back-up ring protect from corrosion and galling
These compact valves are built for maximum durability and robust performance in the toughest applications, and are 100% helium leak-tested to 1 x 104 ml/s for guaranteed performance and reliability. They are available in brass, electroless nickel-plated steel, and electropolished 316 stainless steel. The body-to-bonnet, metal-to-metal seal is designed to significantly increase the pressure range of the valveMore Product Information