LEVIFLOW® Single-Use Ultrasonic Flowmeter Starter Kit
Lieferant: Avantor Fluid Handling
24500-07
MFLX24500-07EA 3330 EUR
MFLX24500-07
LEVIFLOW® Single-Use Ultrasonic Flowmeter Starter Kit
Fluidsensoren
Obtain flow measurements of high-purity fluids.
  • Order the starter kit and a flow sensor to build a complete system
  • Choose from sterile or non-sterile flow sensors covering a range of 0.08 to 800 L/min
  • Avoid contamination— systems don't contain moving or obtrusive parts
  • Order starter bundle for immediate use
These ultrasonic flowmeter systems are designed for use in applications were cleanliness is required. Readings are accurate to 1%—high precision for high-purity fluids. Each system requires a starter kit and clamp-on flow sensor; order separately. The starter kit includes components to get a system in place, just choose a flow sensor at the desired flow range and sterility.
Starter kit includes a single-channel flow converter with DSP for processing the sensor’s signals, connection cables for meter to PC and meter to flow sensor, power supply and cord, and configuration software on a USB stick. The converter also features a 4-digit LED display indicating the flow value.
Flow sensors feature integral sanitary Tri-Clamp® connections and are designed for one-time use. Sensors are IP65 rated and have a watertight connection box.
Order the starter bundle, which includes the starter kit and the ultrasonic sensor, for your convenience.