CPC® Schnell trennbare Fittings für hohen Durchfluss, mit Schlauchnippeleinsätzen
Lieferant: Avantor Fluid Handling
31303-43 31303-46 31303-43BLK 31303-46BLK
MFLX31303-40EA 11.2 EUR
MFLX31303-40 MFLX31303-41 MFLX31303-42 MFLX31303-45 MFLX31303-47 MFLX31303-48 MFLX31307-40 MFLX31307-41 MFLX31307-45 MFLX31307-46 MFLX31307-48 MFLX31303-43 MFLX31303-46 MFLX31303-43BLK MFLX31303-46BLK MFLX31303-42BLK MFLX31303-47BLK MFLX31303-48BLK MFLX31307-40BLK MFLX31307-41BLK MFLX31307-48BLK MFLX31303-40BLK MFLX31303-41BLK MFLX31303-45BLK MFLX31307-45BLK MFLX31307-46BLK
CPC® Schnell trennbare Fittings für hohen Durchfluss, mit Schlauchnippeleinsätzen
Fittings und Verbinder
Keine Leckagen - Kupplungen mit Ventil sperren automatisch den Durchfluss.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Compact, lightweight couplings feature a high flow and low spillage design
These CPC® high-flow quick-disconnect fittings have flow comparable to many ¹/₂" flow couplings in a ³/₈" body size. An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Valved models have a spring-operated 316 stainless steel valve that seals the flow path when fitting halves are disengaged. All fitting bodies have 316 stainless steel latch springs and EPDM seals.
For a complete coupling, order one fitting body and one insert.