CPC® High-Flow Quick-Disconnect Fittings, GHT (M) Bodies and Inserts
Lieferant: Avantor Fluid Handling
31308-76BLK
MFLX31308-76BLKEA 176 EUR
MFLX31308-76BLK
CPC® High-Flow Quick-Disconnect Fittings, GHT (M) Bodies and Inserts
Fittings und Verbinder
No more leaks—valved couplings automatically shut off flow.
  • Compact, lightweight couplings feature a high flow and low spillage design
  • Constructed of durable polysulfone
These CPC® high-flow quick-disconnect fittings are designed to enable enhanced flow in a 3/8” body size. An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Valved models have a spring-operated 316 stainless steel valve that seals the flow path when fitting halves are disengaged. All fitting bodies have 316 stainless steel latch springs and EPDM seals.
For a complete coupling, order one fitting body and one insert.