Grooving Tool for PFA Compression Fittings
Lieferant: Avantor Fluid Handling
31321-49
MFLX31321-49EA 384 EUR
MFLX31321-49
Grooving Tool for PFA Compression Fittings
Werkzeug zum Herausziehen und Einführen
Get a secure seal at higher pressures.
  • Optimize performance of your PFA Compression Fittings
Improve the sealing capability of PTFE fittings. Fittings withstand pressures to 150 psi when a tool is used (40 psi without tool).