Air/N2 Gas Mass Flow Controllers with Metal bodies
Lieferant: Avantor Fluid Handling
32660-04 32660-06 32660-08 32660-10 32660-12 32660-14 32660-16 32660-19 32660-22 32660-24 32661-00 32661-04 32661-08 32661-12 32661-16 32661-22 32665-10 32666-00
MFLX32660-04EA 1060 EUR
MFLX32660-04 MFLX32660-06 MFLX32660-08 MFLX32660-10 MFLX32660-12 MFLX32660-14 MFLX32660-16 MFLX32660-19 MFLX32660-22 MFLX32660-24 MFLX32661-00 MFLX32661-04 MFLX32661-08 MFLX32661-12 MFLX32661-16 MFLX32661-22 MFLX32665-10 MFLX32666-00
Air/N2 Gas Mass Flow Controllers with Metal bodies
Strömungsschalter
Precisely control flow rates with built-in valve.
  • Proven design uses metal body components for added durability
  • Accumulated flow can be logged using an optional totalizer
  • Includes NIST-traceable certificate
These controllers feature an advanced straight-tube sensor that ensures accurate and repeatable results. Gas flow measurements are unaffected by moderate temperature and pressure variations at the inlet. The meter also allows a four-point calibration across the flow range to improve meter linearity.
The flow rate set point can be established by either a local potentiometer or by a remote 4 to 20 mA or 0 to 5 VDC signal. Output data is sent via a 0 to 5 VDC or 4 to 20 mA signal; an analog-to-RS converter is also available for data collection and analysis on your PC.
All controllers include a detachable 3-1/2-digit LCD that can be tilted up to 90° for easy reading. The display is remote mountable to three feet using extension cable 32662-70. The totalizer option 32650-22 will display total accumulated flow.
The controller is protected from polarity reversal or short circuit by a built-in resettable fuse. Aluminum models have wetted materials of anodized aluminum, brass, and Viton; stainless steel (SS) models have wetted materials of SS and Viton.
Accuracy:
These precalibrated flow controllers operate at inlet pressures between 5 and 60 psi and at gas temperatures between 59 and 77 °F (15 to 25 °C) while maintaining the stated percent full-scale accuracy and linearity. When operating beyond 5 to 60 psi, add ±0.01%/psi full-scale; if operating beyond 59 to 77 °F (15 to 25 °C), add ±0.15%/ °C full-scale.
Lieferumfang: Includes nIST-traceable calibration report supplied by the manufacturer.