John Guest Acetal push-to-connect tube-to-hose barb adapters
Lieferant: Avantor Fluid Handling
34006-80 34006-82 34007-05 34007-06 34007-07 34007-08 34007-09 34007-11 34007-37
MFLX34006-79EA 24.3 EUR
MFLX34006-79 MFLX34006-81 MFLX34006-83 MFLX34007-04 MFLX34006-80 MFLX34006-82 MFLX34007-05 MFLX34007-06 MFLX34007-07 MFLX34007-08 MFLX34007-09 MFLX34007-11 MFLX34007-37
John Guest Acetal push-to-connect tube-to-hose barb adapters
Fittings und Verbinder
No need to buy two fitting halves.
  • Meet NSF and FDA requirements for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semirigid plastic (More Product Information