John Guest acetal push-to-connect 3-way divider
Lieferant: Avantor Fluid Handling
MFLX34007-26EA 161 EUR
MFLX34007-26
John Guest acetal push-to-connect 3-way divider
Fittings und Verbinder
No need to buy two fitting halves.
  • Meet NSF and FDA requirements for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon® or silicone, order soft tube supports.