Female Cam to Male NPT Threaded Couplers, Polypropylene
Lieferant: Avantor Fluid Handling
34020-32
MFLX34020-32EA 18.9 EUR
MFLX34020-32
Female Cam to Male NPT Threaded Couplers, Polypropylene
Fittings und Verbinder
Cam couplers provide quick disconnection from system.
  • All fittings have EPDM gaskets
  • Constructed of durable polypropylene
Cam couplers are ideal for providing a quick-disconnect solution to your pump line.