Union Fittings, NPT(M) Nipple, Straight, PTFE
Lieferant: Avantor Fluid Handling
34030-05
MFLX34030-05EA 77.5 EUR
MFLX34030-05
Union Fittings, NPT(M) Nipple, Straight, PTFE
Fittings und Verbinder
Chemically-inert PTFE handles most aggressive chemicals.
  • Constructed of durable PTFE
Chemically inert, PTFE offers outstanding resistance for any use, especially for critical high-purity applications. When subjected to higher pressure, PTFE as a material has a tendency to creep over time. As a result, these fittings work best at lower operating pressures (110 psi maximum operating pressure at ambient temperature for smaller sizes). All fittings are made from virgin PTFE material.