PTFE Reducing Coupler Fittings, Female NPT
Lieferant: Avantor Fluid Handling
34030-32
MFLX34030-32EA 114 EUR
MFLX34030-32
PTFE Reducing Coupler Fittings, Female NPT
Fittings und Verbinder
Chemically-inert PTFE handles most aggressive chemicals.
  • Constructed of durable PTFE
Chemically inert, PTFE offers outstanding resistance for any use, especially for critical high-purity applications. When subjected to higher pressure, PTFE as a material has a tendency to creep over time. As a result, these fittings work best at lower operating pressures (110 psi maximum operating pressure at ambient temperature for smaller sizes). All fittings are made from virgin PTFE material.