Masterflex® Single-Use Bag Port Disc Opposing Barb, Avantor®
Lieferant: Avantor Fluid Handling
MFLX40331-11EA 50.8 EUR
MFLX40331-11 MFLX40331-12 MFLX40331-13 MFLX40331-14 MFLX40331-15
Masterflex® Single-Use Bag Port Disc Opposing Barb, Avantor®
Fittings und Verbinder
Designed for sample port and dip tube applications.
  • Barb sizes range from 1/8" to 1/2"
  • 2-1/4" disc diameter
Masterflex® Single-Use Bag Ports are engineered to be thermal welded or ultrasonic welded to medical and bioreactor bags of similar material. These materials include polyethylene. These bags can be used to transfer fluid, drug compounds, and cell culture in the biopharmaceudical markets. If material validation is needed, please reach out to our techincal support representatives.