Masterflex® Schlauchadapter, Innen-Luer auf Schlauchnippel, Bulkhead, Nylon, Avantor®
Lieferant: Avantor Fluid Handling
MFLX45502-34EA 21.9 EUR
MFLX45502-34
Masterflex® Schlauchadapter, Innen-Luer auf Schlauchnippel, Bulkhead, Nylon, Avantor®
Fittings und Verbinder
Stellen Sie Ihre Prozessintegrität mit vollständiger Materialrückverfolgbarkeit sicher.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Hose barbs are available from ¹/₁₆" to ¹/₄" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.