Masterflex® Adapteranschlüsse, Luer auf Luer, Winkelstück, Avantor®
Lieferant: Avantor Fluid Handling
MFLX45508-26EA 44.8 EUR
MFLX45508-26 MFLX45508-84
Masterflex® Adapteranschlüsse, Luer auf Luer, Winkelstück, Avantor®
Fittings und Verbinder
Vollständige Materialrückverfolgbarkeit durch Sicherstellung der Prozessintegrität.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
More Product Information