Flux® Drum Pump Tube Sets, Polypropylene
Lieferant: Avantor Fluid Handling
70600-04
MFLX70600-04EA 823 EUR
MFLX70600-04
Flux® Drum Pump Tube Sets, Polypropylene
Pumpen Fass/Transferpumpen
Construct a pump system that meets your specific needs.
  • Polypropylene (PP) sets are an economical choice for pumping dilute acids and low-concentration caustics
Standard, High-Head, and Heavy-Duty Tube Sets feature a sealless design for low maintenance and long life. All tube sets include a hose barb fitting made of the same material as the tube set. Fitting accepts either 3/4" or 1" ID tubing.