Masterflex® Pressure Booster Pump Systems, Avantor®
Lieferant: Avantor Fluid Handling
70794-44 70794-64
MFLX70794-44EA 5070 EUR
MFLX70794-44 MFLX70794-64
Masterflex® Pressure Booster Pump Systems, Avantor®
Pumpen Kolbenpumpen
Pump system automatically adjusts speed to maintain set pressure.
  • Maximum flow range of 25 to 100 GPM│NEMA 1 or NEMA 4 variable frequency drive controller with corresponding main disconnect
  • Three-phase input line voltage (single-phase systems are available)
Pump supply booster systems feature a stainless steel centrifugal pump. System is controlled by the variable frequency drive (VFD) which features an LCD and adjustable pressure range; choose the VFD with either a NEMA 1 or NEMA 4 rated enclosure and main disconnect.
System has a small footprint ofMore Product Information