24 VDC Motor Polypropylene Standard Centrifugal Pumps
Lieferant: Avantor Fluid Handling
72008-20
MFLX72008-20EA 454 EUR
MFLX72008-20
24 VDC Motor Polypropylene Standard Centrifugal Pumps
Pumpen Kreiselpumpen
DC-Powered motors are lightweight and portable – excellent for outdoor sampling and transfer of fluids.
  • Sealless pump is great when used with chemically aggressive water samples — no replacement of dirty seals
  • Compact, brushless design
  • Internal cooling circuit reduces bearing temperature
  • Pinpoint bearings allow for run dry capability up to 8 hours
These magnetic drive pumps feature continuous duty 24 VDC motors. Pumps are ideal for use with chemically aggressive water samples as they are constructed of glass fiber-reinforced polypropylene front and rear casings, Viton® o-ring, and either PPS or PTFE bearings.
Lieferumfang: Includes 6 ft (1.8 m) cord with stripped ends.