Fluoroplastic Enclosed Motor Chemical-Resistance Centrifugal Pumps
Lieferant: Avantor Fluid Handling
72011-00 72011-05 72011-10
MFLX72011-00EA 0 EUR
MFLX72011-00 MFLX72011-05 MFLX72011-10
Fluoroplastic Enclosed Motor Chemical-Resistance Centrifugal Pumps
Pumpen Kreiselpumpen
Twin bearing system handles thrust loads for extended service life.
  • Quiet, energy efficient, and compact
  • Internal cooling circuit reduces bearing temperature
  • Enclosed motor for use in harsh environments
Fluoroplastic pumps with carbon fiber-reinforced ETFE are designed for highly corrosive or high-purity applications.
Lieferumfang: Includes a 6-ft (1.8 m) cord with stripped ends.