Neptune Feed Verification Diaphragm Pump Flow Checkers
Lieferant: Avantor Fluid Handling
73805-00
MFLX73805-00EA 1070 EUR
MFLX73805-00
Neptune Feed Verification Diaphragm Pump Flow Checkers
Durchflussanzeiger
Easily meansure flow of your Feed Verification Diaphragm Pump.
  • Optimize performance of your Neptune Metering Pumps
These flow checkers can be mounted directly onto the discharge end of the pumps. The checker is a simple oval gear meter that measures the pump output and then transmits one pulse for every one ml of flow. It requires 4.5 to 25 VDC at a 20 mA maximum. Pump sold separately.