Warren Rupp Air-Powered Double-Diaphragm PVDF Pumps
Lieferant: Avantor Fluid Handling
74000-21 74000-46 74000-80 74000-82
MFLX74000-21EA 2020 EUR
MFLX74000-21 MFLX74000-46 MFLX74000-80 MFLX74000-82
Warren Rupp Air-Powered Double-Diaphragm PVDF Pumps
Pumpen Druckluftpumpen
Move fluids with viscosities up to 11,000 cp.
  • Air-powered motor eliminates sparking concerns associate with electrical pumps
  • Bolted pump design allows easy access to pump head for maintenance and maintains seal after repeated servicing
  • Non-metallic pumps are ideally suited for highly corrosive fluids intermittent/on-demand, portable low abrasive fluids, and suspended solids
  • Safely handles abrasive fluids and slurries — ideal for high-viscosity and shear sensitive materials
  • Encapsulated muffler provides excellent sound dampening
These pumps can moveMore Product Information